Assay Details
| Gene Symbol: Hacl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053493.1 | NP_445945.1 | 4 - 5 | 411 | 66 | |
| GenBank mRNA | AJ517469.1 | - | 4 - 5 | 411 | 66 | |
| BC078697.1 | - | 4 - 5 | 395 | 7110355 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P076352 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P023869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58935 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P076352 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P023869 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P023869 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P076352 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P076352 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P023869 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P383920 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P383928 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14961 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30654 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10790271 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10790274 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10790264 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10790258 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10790263 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10790270 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10790262 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10790272 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10790261 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10790265 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10790260 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10790268 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10790267 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10790269 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10790273 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10790259 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10790275 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10790266 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10790257 | - | - |
| Rat Genome 230 2.0 Array | 1371012_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1371012_at | - | - |
| Affymetrix HT Rat Focus Array | 1371012_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1371012_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1600016023.rn.1 | TC1600002123.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600016043.rn.1 | TC1600002123.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600016036.rn.1 | TC1600002123.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600016031.rn.1 | TC1600002123.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600016032.rn.1 | TC1600002123.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600016033.rn.1 | TC1600002123.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600016041.rn.1 | TC1600002123.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600016047.rn.1 | TC1600002123.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600016027.rn.1 | TC1600002123.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600016028.rn.1 | TC1600002123.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600016030.rn.1 | TC1600002123.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600016038.rn.1 | TC1600002123.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600016044.rn.1 | TC1600002123.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600016024.rn.1 | TC1600002123.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600016035.rn.1 | TC1600002123.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600016040.rn.1 | TC1600002123.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600016042.rn.1 | TC1600002123.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600016026.rn.1 | TC1600002123.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600016034.rn.1 | TC1600002123.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600016037.rn.1 | TC1600002123.rn.1 | RTA TC |
| Rat Expression Set 230 | 1371012_at | - | - |
| Rat Expression Set 230 | 1371012_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10790257 | - | - |
| Rat Genome U34 Set | rc_AA893239_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20854042 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350232 | - | - |
| RatRef-12 Expression BeadChip | ILMN_49010 | - | - |

Hacl1
Genomic Map