Assay Details
Gene Symbol: Prss12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053504.1 | NP_445956.1 | 5 - 6 | 1329 | 89 | |
GenBank mRNA | AJ311671.2 | - | 5 - 6 | 1329 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P423651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 555968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368900 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417785 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163255 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P423651 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P423651 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P423651 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P423651 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 58855 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 91383 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2404 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 17856 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103723 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10818887 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10818884 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10818891 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10818881 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10818882 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10818890 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10818888 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10818889 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10818886 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10818885 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10818883 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10818880 | - | - |
Affymetrix Rat Clariom S Assay | PSR0200014761.rn.1 | TC0200002128.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10818880 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21740830 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363261 | - | - |
RatRef-12 Expression BeadChip | ILMN_52591 | - | - |
