Assay Details
Gene Symbol: Asah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053646.1 | NP_446098.1 | 10 - 11 | 1237 | 77 | |
XM_006231261.2 | XP_006231323.1 | 13 - 14 | 1595 | 77 | ||
XM_006231263.1 | XP_006231325.1 | 12 - 13 | 2173 | 77 | ||
XM_006231267.2 | XP_006231329.1 | 12 - 13 | 2179 | 77 | ||
XM_006231268.2 | XP_006231330.1 | 12 - 13 | 2176 | 77 | ||
XM_006231270.1 | XP_006231332.1 | 12 - 13 | 2176 | 77 | ||
XM_008760319.1 | XP_008758541.1 | 15 - 16 | 4015 | 77 | ||
XM_008760320.1 | XP_008758542.1 | 12 - 13 | 1456 | 77 | ||
GenBank mRNA | AB057433.1 | - | 10 - 11 | 1237 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P102483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506605 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133371 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232473 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40507 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P102483 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P102483 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P102483 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12884 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3426 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 74741 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 87828 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10729704 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729714 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729697 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729698 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729702 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729713 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729701 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729708 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729694 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729703 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729706 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729712 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729707 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729709 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729700 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729695 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729705 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729696 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729699 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729710 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729711 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10729693 | - | - |
Rat Genome 230 2.0 Array | 1369406_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369406_at | - | - |
Affymetrix HT Rat Focus Array | 1369406_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369406_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100060760.rn.1 | TC0100007572.rn.1 | RTA TC |
Rat Expression Set 230 | 1369406_at | - | - |
Rat Expression Set 230 | 1369406_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10729693 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21304577 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363356 | - | - |
RatRef-12 Expression BeadChip | ILMN_49089 | - | - |
