Assay Details
| Gene Symbol: Cdc42bpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053657.1 | NP_446109.1 | 34 - 35 | 5319 | 76 | |
| GenBank mRNA | AF021935.1 | - | 34 - 35 | 5319 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
protein phosphorylation
cytoskeleton organization
nuclear migration
cell migration
actomyosin structure organization
actin cytoskeleton reorganization
intracellular signal transduction
regulation of catalytic activity
regulation of small GTPase mediated signal transduction
magnesium ion binding
protein serine/threonine kinase activity
protein binding
ATP binding
identical protein binding
microtubule cytoskeleton organization
protein phosphorylation
cytoskeleton organization
nuclear migration
cell migration
actomyosin structure organization
actin cytoskeleton reorganization
intracellular signal transduction
regulation of catalytic activity
regulation of small GTPase mediated signal transduction
magnesium ion binding
protein serine/threonine kinase activity
protein binding
ATP binding
identical protein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P138654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11651 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414027 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213599 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83458 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176919 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99857 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546820 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337116 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99200 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P138654 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P138654 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P138654 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P366832 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P529269 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701716702 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 88682 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 56790 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18364 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14366 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 39025 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 61132 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 72684 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101117 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31669 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33421 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 3387 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76896 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 72859 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68051 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13699 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10987 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 104953 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30564 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35133 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 24843 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701716702 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10766154 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766141 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766132 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766128 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766155 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766127 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766144 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766135 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766159 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766129 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766143 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766148 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766152 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766156 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766160 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766124 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766158 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766138 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766131 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766147 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766130 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766145 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766133 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766136 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766157 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766151 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766146 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766125 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766149 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766153 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766142 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766150 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766126 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766140 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766139 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766134 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10766123 | - | - |
| Rat Genome 230 2.0 Array | 1369040_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369040_at | - | - |
| Affymetrix HT Rat Focus Array | 1369040_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369040_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300006320.rn.1 | TC1300000897.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300006319.rn.1 | TC1300000897.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369040_at | - | - |
| Rat Expression Set 230 | 1369040_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766123 | - | - |
| Rat Genome U34 Set | AF021935_at | - | - |
| Rat Genome U34 Set | rc_AI069982_s_at | - | - |
| Rat Genome U34 Set | rc_AI069982_s_at | - | - |
| Rat Genome U34 Set | AF021935_at | - | - |
| Rat Genome U34 Set | AF021935_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21447146 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356750 | - | - |
| RatRef-12 Expression BeadChip | ILMN_62193 | - | - |

Cdc42bpa
Genomic Map