Assay Details
| Gene Symbol: Dusp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053769.3 | NP_446221.2 | 1 - 2 | 511 | 59 | |
| GenBank mRNA | U02553.1 | - | 1 - 2 | 497 | 59 | |
| X84004.1 | - | 1 - 2 | 534 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dusp1
Genomic Map