Assay Details
| Gene Symbol: Dars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053799.2 | NP_446251.1 | 11 - 12 | 1104 | 69 | |
| XM_006249684.2 | XP_006249746.1 | 11 - 12 | 1180 | 69 | ||
| GenBank mRNA | BC072534.1 | - | 11 - 12 | 1229 | 7098427 | 69 |
| J04487.1 | - | 11 - 12 | 1105 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P078528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708908 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703171 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14697 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55381 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P078528 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P078528 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P078528 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P507274 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700483604 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 60505 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103043 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30400 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17206 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 98062 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700483604 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10767369 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767366 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767365 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767370 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767358 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767359 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767360 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767367 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767363 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767362 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767361 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767364 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767357 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10767356 | - | - |
| Rat Genome 230 2.0 Array | 1398795_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1398795_at | - | - |
| Affymetrix HT Rat Focus Array | 1398795_at | - | - |
| Affymetrix HT Rat Focus Array | 1398795_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300008851.rn.1 | TC1300001349.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300008852.rn.1 | TC1300001349.rn.1 | RTA TC |
| Rat Expression Set 230 | 1398795_at | - | - |
| Rat Expression Set 230 | 1398795_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10767356 | - | - |
| Rat Genome U34 Set | U30485mRNA_s_at | - | - |
| Rat Genome U34 Set | rc_AI009682_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21558495 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365101 | - | - |
| RatRef-12 Expression BeadChip | ILMN_58816 | - | - |

Dars
Genomic Map