Assay Details
| Gene Symbol: Slc28a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053863.1 | NP_446315.1 | 6 - 7 | 761 | 67 | |
| XM_006229422.2 | XP_006229484.1 | 8 - 9 | 1786 | 67 | ||
| XM_006229423.2 | XP_006229485.1 | 8 - 9 | 1898 | 67 | ||
| GenBank mRNA | U10279.1 | - | 6 - 7 | 761 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
purine nucleobase transport
pyrimidine nucleobase transport
pyrimidine-containing compound transmembrane transport
nucleoside transmembrane transport
nucleoside binding
pyrimidine nucleobase transmembrane transporter activity
nucleoside:sodium symporter activity
pyrimidine- and adenine-specific:sodium symporter activity
purine nucleobase transport
pyrimidine nucleobase transport
pyrimidine-containing compound transmembrane transport
nucleoside transmembrane transport
nucleoside binding
pyrimidine nucleobase transmembrane transporter activity
nucleoside:sodium symporter activity
pyrimidine- and adenine-specific:sodium symporter activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P091758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892403 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834061 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131061 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119524 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P091758 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P091758 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P091758 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15483 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701833258 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 34552 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10752 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40370 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 94375 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701833258 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10708277 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708266 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708278 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708274 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708272 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708263 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708270 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708279 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708267 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708280 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708273 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708265 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708276 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708269 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708268 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708271 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708264 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708275 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10708262 | - | - |
| Rat Genome 230 2.0 Array | 1369250_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369250_at | - | - |
| Affymetrix HT Rat Focus Array | 1369250_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369250_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100014805.rn.1 | TC0100001880.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100014804.rn.1 | TC0100001880.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369250_at | - | - |
| Rat Expression Set 230 | 1369250_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708262 | - | - |
| Rat Genome U34 Set | U10279_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21986569 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373981 | - | - |
| RatRef-12 Expression BeadChip | ILMN_64335 | - | - |

Slc28a1
Genomic Map