Assay Details
Gene Symbol: Nln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053970.2 | NP_446422.2 | 11 - 12 | 1881 | 86 | |
XM_006231880.2 | XP_006231942.1 | 11 - 12 | 2035 | 86 | ||
XM_006231881.2 | XP_006231943.1 | 12 - 13 | 2113 | 86 | ||
GenBank mRNA | BC072687.1 | - | 11 - 12 | 1881 | 7099208 | 86 |
FQ228111.1 | - | 4 - 5 | 588 | 86 | ||
FQ230641.1 | - | 4 - 5 | 588 | 86 | ||
X87157.1 | - | 11 - 12 | 1989 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P271643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206638 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2613 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P271643 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P271643 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P271643 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P271643 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700813318 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 44347 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62615 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66176 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71646 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 34812 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 17168 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 77930 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700813318 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10821122 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821119 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821127 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821130 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821131 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821132 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821125 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821123 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821118 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821129 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821120 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821133 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821126 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821128 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10821117 | - | - |
Rat Genome 230 2.0 Array | 1382777_PM_at | - | - |
Rat Genome 230 2.0 Array | 1384242_at | - | - |
Rat Genome 230 2.0 Array | 1382777_at | - | - |
Rat Genome 230 2.0 Array | 1369669_at | - | - |
Rat Genome 230 2.0 Array | 1384242_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369669_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369669_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369669_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200019764.rn.1 | TC0200002789.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200019763.rn.1 | TC0200002789.rn.1 | RTA TC |
Rat Expression Set 230 | 1369669_at | - | - |
Rat Expression Set 230 | 1369669_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821117 | - | - |
Rat Genome U34 Set | X87157_at | - | - |
Rat Genome U34 Set | X87157_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22189636 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361076 | - | - |
RatRef-12 Expression BeadChip | ILMN_54505 | - | - |
