Assay Details
Gene Symbol: Cdh17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053977.2 | NP_446429.1 | 5 - 6 | 525 | 70 | |
XM_008763521.1 | XP_008761743.1 | 5 - 6 | 1979 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
oligopeptide transport
homophilic cell adhesion via plasma membrane adhesion molecules
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
oligopeptide transmembrane transport
proton-dependent oligopeptide secondary active transmembrane transporter activity
calcium ion binding
oligopeptide transport
homophilic cell adhesion via plasma membrane adhesion molecules
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
oligopeptide transmembrane transport
proton-dependent oligopeptide secondary active transmembrane transporter activity
calcium ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P13004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214065 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719222 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32232 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P13004 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P13004 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P13004 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13004 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P132825 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 101363 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59799 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 39097 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102606 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10867669 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867672 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867673 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867668 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867674 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867683 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867676 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867677 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867671 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867680 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867678 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867681 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867684 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867682 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867675 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867679 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867685 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867670 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10867667 | - | - |
Rat Genome 230 2.0 Array | 1369224_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369224_at | - | - |
Affymetrix HT Rat Focus Array | 1369224_at | - | - |
Affymetrix HT Rat Focus Array | 1369224_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500001471.rn.1 | TC0500000218.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500001472.rn.1 | TC0500000218.rn.1 | RTA TC |
Rat Expression Set 230 | 1369224_at | - | - |
Rat Expression Set 230 | 1369224_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867667 | - | - |
Rat Genome U34 Set | L46874_s_at | - | - |
Rat Genome U34 Set | X78997_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21480208 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361578 | - | - |
RatRef-12 Expression BeadChip | ILMN_48509 | - | - |
