Assay Details
| Gene Symbol: Ptpn12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057115.2 | NP_476456.2 | 8 - 9 | 767 | 76 | |
| XM_006235868.2 | XP_006235930.1 | 8 - 9 | 818 | 76 | ||
| XM_006235870.1 | XP_006235932.1 | 7 - 8 | 563 | 76 | ||
| XM_008762631.1 | XP_008760853.1 | 6 - 7 | 2977 | 76 | ||
| GenBank mRNA | D38072.1 | - | 8 - 9 | 832 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P002621 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P088930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771458 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755125 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94799 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P088930 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P002621 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P088930 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P002621 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P002621 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P088930 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11397 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19164 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 95912 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74000 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10853192 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853180 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853181 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853187 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853174 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853182 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853176 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853183 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853172 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853179 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853188 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853173 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853190 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853191 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853175 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853178 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853185 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853189 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853184 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853186 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853177 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10853171 | - | - |
| Rat Genome 230 2.0 Array | 1369496_at | - | - |
| Rat Genome 230 2.0 Array | 1369496_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382154_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382154_at | - | - |
| Affymetrix HT Rat Focus Array | 1382154_at | - | - |
| Affymetrix HT Rat Focus Array | 1382154_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369496_at | - | - |
| Affymetrix HT Rat Focus Array | 1369496_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400001302.rn.1 | TC0400000167.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369496_PM_at | - | - |
| Rat Expression Set 230 | 1369496_at | - | - |
| Rat Expression Set 230 | 1382154_PM_at | - | - |
| Rat Expression Set 230 | 1382154_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10853171 | - | - |
| Rat Genome U34 Set | D38072_at | - | - |
| Rat Genome U34 Set | rc_AI012379_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22289377 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376741 | - | - |

Ptpn12
Genomic Map