Assay Details
Gene Symbol: Oxr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197332.1 | NP_001184261.1 | 5 - 6 | 623 | 103 | |
NM_001197907.1 | NP_001184836.1 | 16 - 17 | 2719 | 103 | ||
NM_057153.3 | NP_476494.2 | 13 - 14 | 2320 | 103 | ||
GenBank mRNA | AF333986.1 | - | 5 - 6 | 562 | 103 | |
BC081744.1 | - | 5 - 6 | 605 | 103 | ||
BC166763.1 | - | 13 - 14 | 2536 | 7931847 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P527134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487452 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153552 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28633 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P527134 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P527134 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P527134 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P200146 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P527134 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P263957 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 76909 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 89271 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10896370 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10896375 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10896361 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10896371 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10896363 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10896372 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10896355 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10896369 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10896358 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10896359 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10896364 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10896362 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10896357 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10896374 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10896373 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10896360 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10896365 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10896353 | - | - |
Rat Genome 230 2.0 Array | 1367869_PM_at | - | - |
Rat Genome 230 2.0 Array | 1367869_at | - | - |
Affymetrix HT Rat Focus Array | 1367869_at | - | - |
Affymetrix HT Rat Focus Array | 1367869_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700007823.rn.1 | TC0700001041.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700007824.rn.1 | TC0700001041.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700007821.rn.1 | TC0700001041.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700007826.rn.1 | TC0700001041.rn.1 | RTA TC |
Rat Expression Set 230 | 1367869_at | - | - |
Rat Expression Set 230 | 1367869_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10896353 | - | - |
Rat Genome U34 Set | rc_H33461_at | - | - |
Rat Genome U34 Set | rc_AI012410_at | - | - |
Rat Genome U34 Set | rc_AI045087_at | - | - |
Rat Genome U34 Set | rc_AI171566_at | - | - |
Rat Genome U34 Set | H33462_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21613735 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350714 | - | - |
