Assay Details
Gene Symbol: Smu1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057195.1 | NP_476543.1 | 3 - 4 | 419 | 68 | |
GenBank mRNA | AY029526.1 | - | 3 - 4 | 419 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1016350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133183 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136012 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1016350 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1016350 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1016350 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P156247 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1016350 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 76335 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 38573 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 78349 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10876036 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876044 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876038 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876037 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876039 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876043 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876041 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876033 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876035 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876034 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876042 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876040 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10876032 | - | - |
Rat Genome 230 2.0 Array | 1367935_PM_at | - | - |
Rat Genome 230 2.0 Array | 1367935_at | - | - |
Affymetrix HT Rat Focus Array | 1367935_at | - | - |
Affymetrix HT Rat Focus Array | 1367935_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500018760.rn.1 | TC0500002555.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500018761.rn.1 | TC0500002555.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500018763.rn.1 | TC0500002555.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500018764.rn.1 | TC0500002555.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500018766.rn.1 | TC0500002555.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500018755.rn.1 | TC0500002555.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500018750.rn.1 | TC0500002555.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500018754.rn.1 | TC0500002555.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500018752.rn.1 | TC0500002555.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500018756.rn.1 | TC0500002555.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500018762.rn.1 | TC0500002555.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500018765.rn.1 | TC0500002555.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500018758.rn.1 | TC0500002555.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500018751.rn.1 | TC0500002555.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500018759.rn.1 | TC0500002555.rn.1 | RTA TC |
Rat Expression Set 230 | 1367935_PM_at | - | - |
Rat Expression Set 230 | 1367935_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876032 | - | - |
Rat Genome U34 Set | rc_AA851226_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22423918 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373960 | - | - |
RatRef-12 Expression BeadChip | ILMN_52070 | - | - |
