Assay Details
| Gene Symbol: Ackr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008766664.1 | XP_008764886.1 | 2 - 3 | 774 | 55 | |
| XM_008766665.1 | XP_008764887.1 | 2 - 3 | 1115 | 55 | ||
| GenBank mRNA | U92803.1 | - | 1 - 2 | 122 | 55 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the U92803.1, XM_008766664.1, XM_008766665.1 transcripts.
Set Membership: |
|

Ackr2
Genomic Map