Assay Details
| Gene Symbol: Abcb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080582.1 | NP_542149.1 | 5 - 6 | 1306 | 119 | |
| GenBank mRNA | AJ003004.1 | - | 5 - 6 | 1306 | 119 | |
| BC085712.1 | - | 5 - 6 | 1322 | 7190755 | 119 | |
| FQ211570.1 | - | 5 - 6 | 1332 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
porphyrin-containing compound biosynthetic process
transport
brain development
heme transport
skin development
transmembrane transport
ATP binding
heme-transporting ATPase activity
efflux transmembrane transporter activity
heme binding
ATPase activity, coupled to transmembrane movement of substances
porphyrin-containing compound biosynthetic process
transport
brain development
heme transport
skin development
transmembrane transport
ATP binding
heme-transporting ATPase activity
efflux transmembrane transporter activity
heme binding
ATPase activity, coupled to transmembrane movement of substances
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P090752 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P446882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146440 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P446882 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P090752 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P446882 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P090752 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P446882 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P090752 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P446882 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19621 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5079 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10929076 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929067 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929080 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929073 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929074 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929070 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929065 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929081 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929083 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929085 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929069 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929064 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929075 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929072 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929084 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929071 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929068 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929066 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929078 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929079 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929082 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929077 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10929063 | - | - |
| Rat Genome 230 2.0 Array | 1382807_at | - | - |
| Rat Genome 230 2.0 Array | 1384261_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368159_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368159_at | - | - |
| Rat Genome 230 2.0 Array | 1382807_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1384261_at | - | - |
| Affymetrix HT Rat Focus Array | 1368159_at | - | - |
| Affymetrix HT Rat Focus Array | 1368159_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0900018453.rn.1 | TC0900002261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900018459.rn.1 | TC0900002261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900018469.rn.1 | TC0900002261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900018448.rn.1 | TC0900002261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900018463.rn.1 | TC0900002261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900018465.rn.1 | TC0900002261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900018461.rn.1 | TC0900002261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900018471.rn.1 | TC0900002261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900018451.rn.1 | TC0900002261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900018455.rn.1 | TC0900002261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900018467.rn.1 | TC0900002261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900018456.rn.1 | TC0900002261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900018458.rn.1 | TC0900002261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900018466.rn.1 | TC0900002261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900018449.rn.1 | TC0900002261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900018457.rn.1 | TC0900002261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900018470.rn.1 | TC0900002261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900018454.rn.1 | TC0900002261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900018460.rn.1 | TC0900002261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900018468.rn.1 | TC0900002261.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368159_at | - | - |
| Rat Expression Set 230 | 1368159_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10929063 | - | - |
| Rat Genome U34 Set | AF106563_s_at | - | - |
| Rat Genome U34 Set | AF106563_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22026401 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355792 | - | - |
| RatRef-12 Expression BeadChip | ILMN_59060 | - | - |

Abcb6
Genomic Map