Assay Details
| Gene Symbol: Cacng3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080691.1 | NP_542422.1 | 2 - 3 | 293 | 95 | |
| GenBank mRNA | AF361340.1 | - | 2 - 3 | 293 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P129720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693599 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508844 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49519 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158057 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P129720 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P129720 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P129720 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P341847 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30659 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55362 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80443 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80549 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103339 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 24177 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10710671 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710670 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710673 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710672 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10710669 | - | - |
| Rat Genome 230 2.0 Array | 1370757_at | - | - |
| Rat Genome 230 2.0 Array | 1370757_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370757_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370757_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100019606.rn.1 | TC0100002538.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100019605.rn.1 | TC0100002538.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370757_PM_at | - | - |
| Rat Expression Set 230 | 1370757_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710669 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21615504 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1366333 | - | - |
| RatRef-12 Expression BeadChip | ILMN_66354 | - | - |

Cacng3
Genomic Map