Assay Details
| Gene Symbol: Afap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080900.1 | NP_543176.1 | 12 - 13 | 1655 | 99 | |
| XM_008770245.1 | XP_008768467.1 | 13 - 14 | 1814 | 99 | ||
| XM_008770246.1 | XP_008768468.1 | 13 - 14 | 1814 | 99 | ||
| XM_008770247.1 | XP_008768469.1 | 14 - 15 | 2084 | 99 | ||
| GenBank mRNA | AY063759.1 | - | 12 - 13 | 1655 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P207963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70032 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P207963 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P207963 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P207963 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P207963 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 64586 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 52885 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22239 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43991 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 53829 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 62006 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10777458 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777464 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777459 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777465 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777462 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777460 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777453 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777456 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777451 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777463 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777457 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777454 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777461 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777466 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777452 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777467 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777455 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10777450 | - | - |
| Rat Genome 230 2.0 Array | 1369385_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1381177_at | - | - |
| Rat Genome 230 2.0 Array | 1381177_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369385_at | - | - |
| Affymetrix HT Rat Focus Array | 1369385_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369385_at | - | - |
| Affymetrix HT Rat Focus Array | 1381177_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1381177_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1400014645.rn.1 | TC1400002061.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400014646.rn.1 | TC1400002061.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400014637.rn.1 | TC1400002061.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400014642.rn.1 | TC1400002061.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400014644.rn.1 | TC1400002061.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400014648.rn.1 | TC1400002061.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400014647.rn.1 | TC1400002061.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400014631.rn.1 | TC1400002061.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400014641.rn.1 | TC1400002061.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400014628.rn.1 | TC1400002061.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400014635.rn.1 | TC1400002061.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400014630.rn.1 | TC1400002061.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400014634.rn.1 | TC1400002061.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400014638.rn.1 | TC1400002061.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400014643.rn.1 | TC1400002061.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400014636.rn.1 | TC1400002061.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400014640.rn.1 | TC1400002061.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400014649.rn.1 | TC1400002061.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400014650.rn.1 | TC1400002061.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369385_at | - | - |
| Rat Expression Set 230 | 1369385_PM_at | - | - |
| Rat Expression Set 230 | 1381177_PM_at | - | - |
| Rat Expression Set 230 | 1381177_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10777450 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21726283 | - | - |

Afap1
Genomic Map