Assay Details
Gene Symbol: Slc28a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080908.1 | NP_543184.1 | 5 - 6 | 574 | 65 | |
XM_008771422.1 | XP_008769644.1 | 5 - 6 | 659 | 65 | ||
GenBank mRNA | AY059414.1 | - | 5 - 6 | 574 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
retina homeostasis
purine nucleobase transport
pyrimidine nucleobase transport
purine nucleoside transmembrane transport
pyrimidine nucleoside transport
sodium ion transmembrane transport
pyrimidine-containing compound transmembrane transport
nucleoside transmembrane transport
nucleoside binding
nucleoside:sodium symporter activity
pyrimidine- and adenine-specific:sodium symporter activity
purine-specific nucleoside:sodium symporter activity
retina homeostasis
purine nucleobase transport
pyrimidine nucleobase transport
purine nucleoside transmembrane transport
pyrimidine nucleoside transport
sodium ion transmembrane transport
pyrimidine-containing compound transmembrane transport
nucleoside transmembrane transport
nucleoside binding
nucleoside:sodium symporter activity
pyrimidine- and adenine-specific:sodium symporter activity
purine-specific nucleoside:sodium symporter activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P012749 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369443 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83450 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16457 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P012749 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P012749 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P012749 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13100 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73816 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 54981 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48035 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10793803 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793802 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793810 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793814 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793801 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793817 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793813 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793807 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793804 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793809 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793805 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793811 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793812 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793815 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793808 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793816 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793806 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793800 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10793799 | - | - |
Rat Genome 230 2.0 Array | 1369447_at | - | - |
Rat Genome 230 2.0 Array | 1369447_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369447_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369447_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1700000694.rn.1 | TC1700000100.rn.1 | RTA TC |
Rat Expression Set 230 | 1369447_at | - | - |
Rat Expression Set 230 | 1369447_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10793799 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21563347 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369304 | - | - |
RatRef-12 Expression BeadChip | ILMN_54330 | - | - |
