Assay Details
| Gene Symbol: Abcg8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130414.2 | NP_569098.2 | 6 - 7 | 1071 | 66 | |
| XM_008764453.1 | XP_008762675.1 | 4 - 5 | 1112 | 66 | ||
| GenBank mRNA | AF351785.2 | - | 6 - 7 | 1071 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcg8
Genomic Map