Assay Details
| Gene Symbol: Gucy2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130737.4 | NP_570093.1 | 18 - 19 | 3359 | 73 | |
| XM_006229710.1 | XP_006229772.1 | 18 - 19 | 3358 | 73 | ||
| GenBank mRNA | L37203.1 | - | 18 - 19 | 3359 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P105402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583667 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129696 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P105402 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P105402 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P105402 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P13122 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 21569 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 32271 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10708878 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708895 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708889 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708883 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708892 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708876 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708893 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708884 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708890 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708879 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708888 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708881 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708877 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708894 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708882 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708886 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708885 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708891 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708880 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708887 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10708874 | - | - |
| Rat Genome 230 2.0 Array | 1369223_at | - | - |
| Rat Genome 230 2.0 Array | 1369223_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369223_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369223_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100016124.rn.1 | TC0100002094.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100016123.rn.1 | TC0100002094.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369223_at | - | - |
| Rat Expression Set 230 | 1369223_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708874 | - | - |
| Rat Genome U34 Set | L37203ExpandedCDS_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21570631 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352322 | - | - |
| RatRef-12 Expression BeadChip | ILMN_60310 | - | - |

Gucy2d
Genomic Map