Assay Details
Gene Symbol: Kcnj15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133321.2 | NP_579855.1 | 1 - 2 | 101 | 68 | |
GenBank mRNA | AY028455.1 | - | 1 - 2 | 102 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AY028455.1, NM_133321.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
