Assay Details
| Gene Symbol: Il4r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133380.2 | NP_596871.2 | 1 - 2 | 69 | 98 | |
| GenBank mRNA | AB015746.1 | - | 1 - 2 | 76 | 98 | |
| AB015747.1 | - | 1 - 2 | 76 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB015746.1, AB015747.1, NM_133380.2 transcripts.
Set Membership: |
|

Il4r
Genomic Map