Assay Details
Gene Symbol: Sstr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133522.1 | NP_598206.1 | 2 - 3 | 613 | 62 | |
GenBank mRNA | X63574.1 | - | 2 - 3 | 613 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_133522.1, X63574.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
