Assay Details
Gene Symbol: Ccr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133532.2 | NP_598216.2 | 1 | 953 | 85 | |
XM_006243915.2 | XP_006243977.1 | 2 | 1140 | 85 | ||
GenBank mRNA | AF432872.2 | - | 1 | 953 | 85 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
