Assay Details
| Gene Symbol: Rab3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133536.2 | NP_598220.1 | 1 - 2 | 108 | 92 | |
| GenBank mRNA | D78197.1 | - | 1 - 2 | 85 | 92 | |
| U54807.1 | - | 1 - 2 | 112 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the D78197.1, NM_133536.2, U54807.1 transcripts.
Set Membership: |
|

Rab3c
Genomic Map