Assay Details
Gene Symbol: Itfg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133557.2 | NP_598241.2 | 7 - 8 | 859 | 105 | |
GenBank mRNA | AF480856.2 | - | 7 - 8 | 735 | 105 | |
BC085706.1 | - | 7 - 8 | 859 | 7189102 | 105 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P784252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46507 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105975 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P784252 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P784252 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P784252 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P784252 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 85552 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 34014 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53054 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72004 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2411 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 70321 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37632 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72616 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 101700 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10806428 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806422 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806419 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806423 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806421 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806425 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806420 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806427 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806424 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806426 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806418 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806433 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806429 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806432 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806431 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806434 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806417 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806430 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806435 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10806416 | - | - |
Rat Genome 230 2.0 Array | 1367716_at | - | - |
Rat Genome 230 2.0 Array | 1367716_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1367716_at | - | - |
Affymetrix HT Rat Focus Array | 1367716_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1900001896.rn.1 | TC1900000253.rn.1 | RTA TC |
Rat Expression Set 230 | 1367716_at | - | - |
Rat Expression Set 230 | 1367716_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806416 | - | - |
Rat Genome U34 Set | rc_AI230037_at | - | - |
Rat Genome U34 Set | rc_AI102032_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20743259 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376361 | - | - |
