Assay Details
| Gene Symbol: Lgals2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133599.1 | NP_598283.1 | 1 - 2 | 43 | 127 | |
| GenBank mRNA | AB001075.1 | - | 1 - 2 | 43 | 127 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB001075.1, NM_133599.1 transcripts.
Set Membership: |
|

Lgals2
Genomic Map