Assay Details
| Gene Symbol: Ppp1r3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006253215.2 | XP_006253277.1 | 1 - 2 | 769 | 97 | |
| GenBank mRNA | Y18208.1 | - | 1 - 2 | 66 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006253215.2, Y18208.1 transcripts.
Set Membership: |
|

Ppp1r3b
Genomic Map