Assay Details
| Gene Symbol: Dmgdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139102.2 | NP_620802.2 | 6 - 7 | 1006 | 76 | |
| XM_008760629.1 | XP_008758851.1 | 6 - 7 | 1021 | 76 | ||
| XM_008760630.1 | XP_008758852.1 | 6 - 7 | 1021 | 76 | ||
| XM_008760631.1 | XP_008758853.1 | 6 - 7 | 1021 | 76 | ||
| XM_008760632.1 | XP_008758854.1 | 6 - 7 | 1021 | 76 | ||
| GenBank mRNA | BC085697.1 | - | 6 - 7 | 1006 | 7108858 | 76 |
| X55995.1 | - | 6 - 7 | 974 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
glycine metabolic process
amino-acid betaine catabolic process
choline metabolic process
tetrahydrofolate interconversion
choline catabolic process
oxidation-reduction process
folic acid binding
poly(A) RNA binding
dimethylglycine dehydrogenase activity
flavin adenine dinucleotide binding
glycine metabolic process
amino-acid betaine catabolic process
choline metabolic process
tetrahydrofolate interconversion
choline catabolic process
oxidation-reduction process
folic acid binding
poly(A) RNA binding
dimethylglycine dehydrogenase activity
flavin adenine dinucleotide binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P454172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303365 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623957 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502528 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160725 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P454172 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P454172 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P454172 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P454172 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701256537 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 61393 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 60891 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5345 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 16215 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101844 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 16017 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63007 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701256537 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10812505 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10812514 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10812508 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10812504 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10812511 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10812512 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10812503 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10812501 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10812506 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10812509 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10812513 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10812510 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10812500 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10812507 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10812502 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10812499 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10812498 | - | - |
| Rat Genome 230 2.0 Array | 1370936_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370936_at | - | - |
| Affymetrix HT Rat Focus Array | 1370936_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370936_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200000845.rn.1 | TC0200000167.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370936_PM_at | - | - |
| Rat Expression Set 230 | 1370936_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10812498 | - | - |
| Rat Genome U34 Set | rc_AA892345_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20713880 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364121 | - | - |
| RatRef-12 Expression BeadChip | ILMN_48687 | - | - |

Dmgdh
Genomic Map