Assay Details
Gene Symbol: Fgd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139263.1 | NP_640356.1 | 11 - 12 | 2786 | 141 | |
XM_008768850.1 | XP_008767072.1 | 11 - 12 | 2162 | 141 | ||
GenBank mRNA | AF038388.1 | - | 11 - 12 | 2786 | 141 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -



Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P244257 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P074940 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P031118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520739 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169877 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P244257 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P074940 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P031118 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P074940 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P244257 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P031118 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P031118 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P244257 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P074940 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P244257 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P233887 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3396 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 60187 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 51354 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 33659 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 60783 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 76991 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10755881 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755880 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755889 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755887 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755876 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755877 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755878 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755884 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755879 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755885 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755888 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755883 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755882 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755886 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10755875 | - | - |
Rat Genome 230 2.0 Array | 1370600_PM_at | - | - |
Rat Genome 230 2.0 Array | 1370600_at | - | - |
Affymetrix HT Rat Focus Array | 1370600_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370600_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1100006229.rn.1 | TC1100001034.rn.1 | RTA TC |
Rat Expression Set 230 | 1370600_at | - | - |
Rat Expression Set 230 | 1370600_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755875 | - | - |
Rat Genome U34 Set | AF038388_at | - | - |
Rat Genome U34 Set | AF038388_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22122832 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365078 | - | - |
RatRef-12 Expression BeadChip | ILMN_48421 | - | - |
