Assay Details
Gene Symbol: Tpcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139332.3 | NP_647548.2 | 25 - 26 | 2369 | 64 | |
GenBank mRNA | AB018253.2 | - | 24 - 25 | 2372 | 64 | |
BC062072.1 | - | 24 - 25 | 2369 | 5599545 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
calcium ion transport
biological_process
ion transmembrane transport
regulation of ion transmembrane transport
calcium ion transmembrane transport
membrane depolarization during action potential
molecular_function
voltage-gated calcium channel activity
identical protein binding
NAADP-sensitive calcium-release channel activity
calcium ion transport
biological_process
ion transmembrane transport
regulation of ion transmembrane transport
calcium ion transmembrane transport
membrane depolarization during action potential
molecular_function
voltage-gated calcium channel activity
identical protein binding
NAADP-sensitive calcium-release channel activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P030550 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P170524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607713 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12095 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P170524 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P030550 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P170524 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P030550 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P030550 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P170524 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P170524 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700147464 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 101642 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49048 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 39438 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84054 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700147464 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10758829 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758820 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758814 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758813 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758811 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758815 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758824 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758823 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758809 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758825 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758826 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758828 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758827 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758818 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758817 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758822 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758812 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758819 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758816 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758821 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758810 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10758808 | - | - |
Rat Genome 230 2.0 Array | 1370358_at | - | - |
Rat Genome 230 2.0 Array | 1370358_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370358_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370358_at | - | - |
Rat Expression Set 230 | 1370358_at | - | - |
Rat Expression Set 230 | 1370358_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10758808 | - | - |
Rat Genome U34 Set | rc_AA891614_at | - | - |
Rat Genome U34 Set | rc_AI230042_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21588056 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371451 | - | - |
RatRef-12 Expression BeadChip | ILMN_51591 | - | - |
