Assay Details
| Gene Symbol: Dync1li1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145772.1 | NP_665715.1 | 8 - 9 | 1105 | 147 | |
| GenBank mRNA | AF181992.1 | - | 8 - 9 | 1105 | 147 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P269119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539573 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338729 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120680 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P269119 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P269119 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P269119 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P269119 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701319068 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78742 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10408 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 104971 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701319068 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10914002 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914003 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914006 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10913998 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914000 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914008 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914001 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10913996 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10913999 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914004 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10913997 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914005 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10914007 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10913995 | - | - |
| Rat Genome 230 2.0 Array | 1370576_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374297_at | - | - |
| Rat Genome 230 2.0 Array | 1370576_at | - | - |
| Rat Genome 230 2.0 Array | 1399090_at | - | - |
| Rat Genome 230 2.0 Array | 1399090_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374297_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374297_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370576_at | - | - |
| Affymetrix HT Rat Focus Array | 1370576_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1374297_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800013185.rn.1 | TC0800001660.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370576_PM_at | - | - |
| Rat Expression Set 230 | 1374297_PM_at | - | - |
| Rat Expression Set 230 | 1374297_at | - | - |
| Rat Expression Set 230 | 1399090_PM_at | - | - |
| Rat Expression Set 230 | 1370576_at | - | - |
| Rat Expression Set 230 | 1399090_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10913995 | - | - |
| Rat Genome U34 Set | rc_H31847_at | - | - |
| Rat Genome U34 Set | rc_AI009555_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21896103 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369108 | - | - |
| RatRef-12 Expression BeadChip | ILMN_65087 | - | - |

Dync1li1
Genomic Map