Assay Details
| Gene Symbol: Obp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_147215.2 | NP_671748.2 | 620 | 136 | ||
| GenBank mRNA | J00738.1 | - | 620 | 136 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Obp3
Genomic Map