Assay Details
| Gene Symbol: Prop1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153627.1 | NP_705891.1 | 1 - 2 | 116 | 73 | |
| XM_008767664.1 | XP_008765886.1 | 1 - 2 | 113 | 73 | ||
| GenBank mRNA | AB037922.1 | - | 1 - 2 | 116 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prop1
Genomic Map