Assay Details
| Gene Symbol: Duox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153739.1 | NP_714961.1 | 1 - 2 | 89 | 73 | |
| XM_006234829.2 | XP_006234891.1 | 2 - 3 | 73 | |||
| GenBank mRNA | AF542180.1 | - | 1 - 2 | 89 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
thyroid hormone generation
response to oxidative stress
cytokine-mediated signaling pathway
cuticle development
hormone biosynthetic process
hydrogen peroxide catabolic process
hydrogen peroxide biosynthetic process
response to cAMP
oxidation-reduction process
reactive oxygen species metabolic process
peroxidase activity
calcium ion binding
NAD(P)H oxidase activity
heme binding
thyroid hormone generation
response to oxidative stress
cytokine-mediated signaling pathway
cuticle development
hormone biosynthetic process
hydrogen peroxide catabolic process
hydrogen peroxide biosynthetic process
response to cAMP
oxidation-reduction process
reactive oxygen species metabolic process
peroxidase activity
calcium ion binding
NAD(P)H oxidase activity
heme binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P086118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287441 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6912 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P086118 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P086118 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P086118 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P332432 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30443 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99643 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31222 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46382 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10839270 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839284 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839282 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839280 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839281 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839290 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839279 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839294 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839292 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839277 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839283 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839291 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839288 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839273 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839293 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839295 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839287 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839276 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839286 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839278 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839274 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839271 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839269 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839285 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839272 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10839289 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10839268 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10849230 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300011100.rn.1 | TC0300001589.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300011101.rn.1 | TC0300001589.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10839268 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21138617 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367874 | - | - |

Duox1
Genomic Map