Assay Details
| Gene Symbol: Rims4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170666.1 | NP_733766.1 | 1 - 2 | 103 | 65 | |
| GenBank mRNA | AF548739.1 | - | 1 - 2 | 103 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rims4
Genomic Map