Assay Details
Gene Symbol: Ush2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY077844.1 | - | 11 - 12 | 2339 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P297053 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P297053 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P297053 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P297053 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P245490 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P22165 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P222266 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 18109 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56907 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37569 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73681 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 79534 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2647 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 52844 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10766610 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766603 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766616 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766605 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766614 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766609 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766604 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766618 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766615 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766602 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766613 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766608 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766601 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766619 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766607 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766606 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766617 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10766600 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10766600 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20756403 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369513 | - | - |
