Assay Details
| Gene Symbol: Arhgap39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173122.3 | NP_775145.2 | 11 - 12 | 3363 | 83 | |
| XM_006241866.2 | XP_006241928.1 | 10 - 11 | 3318 | 83 | ||
| XM_006241867.2 | XP_006241929.1 | 10 - 11 | 3308 | 83 | ||
| GenBank mRNA | CB578147.1 | - | 3 - 4 | 460 | 83 | |
| X53232.1 | - | 1 - 2 | 106 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the X53232.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P069405 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P069404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10218 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70202 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147823 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P069405 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P069404 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P069405 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P069404 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P069404 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P069405 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P543694 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700588488 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 24481 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 37001 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 98702 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 21550 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700588488 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10905132 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905130 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905131 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905129 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905140 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905138 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905133 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905135 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905136 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905139 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905137 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905134 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10905128 | - | - |
| Rat Genome 230 2.0 Array | 1388165_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1388165_at | - | - |
| Affymetrix HT Rat Focus Array | 1388165_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1388165_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700026707.rn.1 | TC0700003411.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700026705.rn.1 | TC0700003411.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700026706.rn.1 | TC0700003411.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700026713.rn.1 | TC0700003411.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700026716.rn.1 | TC0700003411.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700026709.rn.1 | TC0700003411.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700026710.rn.1 | TC0700003411.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700026714.rn.1 | TC0700003411.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700026708.rn.1 | TC0700003411.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700026711.rn.1 | TC0700003411.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700026712.rn.1 | TC0700003411.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700026715.rn.1 | TC0700003411.rn.1 | RTA TC |
| Rat Expression Set 230 | 1388165_at | - | - |
| Rat Expression Set 230 | 1388165_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905128 | - | - |
| Rat Genome U34 Set | X53232_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22078064 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365990 | - | - |

Arhgap39
Genomic Map