Assay Details
Gene Symbol: Obp2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173148.1 | NP_775171.1 | 448 | 66 | ||
XM_008761579.1 | XP_008759801.1 | 5 - 6 | 553 | 66 | ||
GenBank mRNA | M76734.1 | - | 448 | 66 |
Gene Symbol: LOC103691751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008761619.1 | XP_008759841.1 | 454 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
