Assay Details
| Gene Symbol: Lgr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173328.1 | NP_775450.1 | 2 - 3 | 301 | 103 | |
| GenBank mRNA | AF061443.1 | - | 2 - 3 | 301 | 103 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lgr4
Genomic Map