Assay Details
| Gene Symbol: Slco6c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173338.1 | NP_775460.1 | 2018 | 149 | ||
| XM_008767361.1 | XP_008765583.1 | 1900 | 149 | |||
| GenBank mRNA | AF326113.1 | - | 2018 | 149 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P332483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 51676 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951786 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178526 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18366 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P332483 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P332483 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P332483 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P332483 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35018 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76087 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1877 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63513 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35438 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 20368 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6549 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10930138 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930142 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930137 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930144 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930135 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930133 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930140 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930143 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930139 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930141 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930145 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930146 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930134 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10930136 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10930132 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0900020844.rn.1 | TC0900002535.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0900020845.rn.1 | TC0900002535.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10930132 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20810619 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361509 | - | - |
| RatRef-12 Expression BeadChip | ILMN_48686 | - | - |

Slco6c1
Genomic Map