Assay Details
| Gene Symbol: Cyp4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044770.2 | NP_001038235.1 | 6 - 7 | 787 | 65 | |
| GenBank mRNA | BC078684.1 | - | 6 - 7 | 787 | 7109305 | 65 |
| BC098705.1 | - | 6 - 7 | 782 | 7106510 | 65 | |
| Gene Symbol: Cyp4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175760.2 | NP_786936.1 | 4 - 5 | 792 | 65 | |
| GenBank mRNA | M33936.1 | - | 4 - 5 | 792 | 65 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
kidney development
response to hormone
arachidonic acid metabolic process
response to drug
linoleic acid metabolic process
icosanoid biosynthetic process
lauric acid metabolic process
oxidation-reduction process
monooxygenase activity
fatty acid binding
iron ion binding
arachidonic acid monooxygenase activity
arachidonic acid 11,12-epoxygenase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
alkane 1-monooxygenase activity
heme binding
arachidonic acid binding
arachidonic acid omega-hydroxylase activity
kidney development
response to hormone
arachidonic acid metabolic process
response to drug
linoleic acid metabolic process
icosanoid biosynthetic process
lauric acid metabolic process
oxidation-reduction process
monooxygenase activity
fatty acid binding
iron ion binding
arachidonic acid monooxygenase activity
arachidonic acid 11,12-epoxygenase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
alkane 1-monooxygenase activity
heme binding
arachidonic acid binding
arachidonic acid omega-hydroxylase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P15784 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P019361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617697 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462977 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285596 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74347 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179397 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P019361 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P15784 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P019361 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P15784 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P15784 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P019361 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15784 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701651574 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 95872 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 9732 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76513 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 16308 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35823 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85604 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30670 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80755 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 97668 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30295 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 9007 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78338 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 81919 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 45230 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701651574 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10878788 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878785 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878786 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878782 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878796 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878791 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878781 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878789 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878790 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878784 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878793 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878783 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878792 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878794 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878795 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10878787 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10878780 | - | - |
| Rat Genome 230 2.0 Array | 1394844_PM_s_at | - | - |
| Rat Genome 230 2.0 Array | 1394844_s_at | - | - |
| Rat Genome 230 2.0 Array | 1370397_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370397_at | - | - |
| Affymetrix HT Rat Focus Array | 1370397_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370397_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500033118.rn.1 | TC0500004261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500033119.rn.1 | TC0500004261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500033112.rn.1 | TC0500004261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500033116.rn.1 | TC0500004261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500033113.rn.1 | TC0500004261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500033115.rn.1 | TC0500004261.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500033117.rn.1 | TC0500004261.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370397_PM_at | - | - |
| Rat Expression Set 230 | 1370397_at | - | - |
| Rat Expression Set 230 | 1394844_PM_s_at | - | - |
| Rat Expression Set 230 | 1394844_s_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10878787 | - | - |
| Rat Genome U34 Set | M33936_s_at | - | - |
| Rat Genome U34 Set | rc_AA924591_at | - | - |
| Rat Genome U34 Set | M33936_s_at | - | - |
| Rat Genome U34 Set | rc_AA924591_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21014073 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364951 | - | - |
| RatRef-12 Expression BeadChip | ILMN_51991 | - | - |

Cyp4a2
Genomic Map