Assay Details
| Gene Symbol: S100vp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176076.3 | NP_788265.1 | 3 - 4 | 505 | 88 | |
| GenBank mRNA | AY195741.1 | - | 3 - 4 | 509 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

S100vp
Genomic Map