Assay Details
| Gene Symbol: Kcnmb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176861.1 | NP_789831.1 | 1 - 2 | 50 | 67 | |
| GenBank mRNA | AY191836.1 | - | 1 - 2 | 50 | 67 | |
| BC081695.1 | - | 350 | 7190722 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P057631 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P154243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459088 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763901 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734400 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908235 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524848 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17072 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542688 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110295 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669176 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32819 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P057631 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P154243 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P154243 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P057631 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P057631 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P154243 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P482475 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 21835 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14328 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 45567 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 79365 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22517 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 822 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19162 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22839 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 9630 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23780 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 37449 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 64343 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15091 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46590 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 89894 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 102462 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 72895 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33191 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 41457 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10814708 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814711 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814712 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814710 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814714 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814709 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814713 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814715 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10814716 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10814707 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200005747.rn.1 | TC0200000948.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200005741.rn.1 | TC0200000948.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200005746.rn.1 | TC0200000948.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200005743.rn.1 | TC0200000948.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200005745.rn.1 | TC0200000948.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200005749.rn.1 | TC0200000948.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200005744.rn.1 | TC0200000948.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200005748.rn.1 | TC0200000948.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10814707 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21663393 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355562 | - | - |
| RatRef-12 Expression BeadChip | ILMN_63088 | - | - |

Kcnmb2
Genomic Map