Assay Details
Gene Symbol: Tmem62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_230497.8 | XP_230497.4 | 2 - 3 | 1101 | 62 | |
GenBank mRNA | FQ210040.1 | - | 2 - 3 | 505 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
