Assay Details
Gene Symbol: Cela2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012553.2 | NP_036685.1 | 6 - 7 | 667 | 71 | |
GenBank mRNA | FQ226773.1 | - | 6 - 7 | 669 | 71 | |
FQ227652.1 | - | 6 - 7 | 669 | 71 | ||
FQ227675.1 | - | 6 - 7 | 669 | 71 | ||
FQ228000.1 | - | 6 - 7 | 669 | 71 | ||
FQ228239.1 | - | 5 - 6 | 550 | 71 | ||
V01233.1 | - | 6 - 7 | 667 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
