Assay Details
| Gene Symbol: Mgmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012861.1 | NP_036993.1 | 2 - 3 | 191 | 148 | |
| GenBank mRNA | M76704.1 | - | 1 - 2 | 136 | 148 | |
| S61804.1 | - | 2 - 3 | 191 | 148 | ||
| X54862.1 | - | 2 - 3 | 217 | 148 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mgmt
Genomic Map