Assay Details
Gene Symbol: Pak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017198.1 | NP_058894.1 | 10 - 11 | 1383 | 98 | |
XM_006229752.1 | XP_006229814.1 | 10 - 11 | 1234 | 98 | ||
XM_006229753.1 | XP_006229815.1 | 10 - 11 | 1231 | 98 | ||
XM_006229754.2 | XP_006229816.1 | 10 - 11 | 1223 | 98 | ||
GenBank mRNA | U23443.1 | - | 10 - 11 | 1189 | 98 | |
U49953.1 | - | 10 - 11 | 1383 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPKK activity
mitotic cell cycle
response to hypoxia
positive regulation of protein phosphorylation
protein phosphorylation
exocytosis
apoptotic process
small GTPase mediated signal transduction
axonogenesis
neuromuscular junction development
response to organic substance
dendrite development
actin cytoskeleton reorganization
cellular response to insulin stimulus
regulation of actin cytoskeleton organization
positive regulation of peptidyl-serine phosphorylation
positive regulation of intracellular estrogen receptor signaling pathway
intracellular signal transduction
wound healing
receptor clustering
positive regulation of JUN kinase activity
protein autophosphorylation
branching morphogenesis of an epithelial tube
neuron projection morphogenesis
positive regulation of stress fiber assembly
negative regulation of cell proliferation involved in contact inhibition
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase kinase activity
protein binding
collagen binding
ATP binding
protein kinase binding
MAPK cascade
activation of MAPKK activity
mitotic cell cycle
response to hypoxia
positive regulation of protein phosphorylation
protein phosphorylation
exocytosis
apoptotic process
small GTPase mediated signal transduction
axonogenesis
neuromuscular junction development
response to organic substance
dendrite development
actin cytoskeleton reorganization
cellular response to insulin stimulus
regulation of actin cytoskeleton organization
positive regulation of peptidyl-serine phosphorylation
positive regulation of intracellular estrogen receptor signaling pathway
intracellular signal transduction
wound healing
receptor clustering
positive regulation of JUN kinase activity
protein autophosphorylation
branching morphogenesis of an epithelial tube
neuron projection morphogenesis
positive regulation of stress fiber assembly
negative regulation of cell proliferation involved in contact inhibition
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase kinase activity
protein binding
collagen binding
ATP binding
protein kinase binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P325005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743785 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359344 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649600 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946749 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161041 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P325005 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P325005 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P325005 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P325005 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701187862 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700499325 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6916 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90830 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 82970 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 91172 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 13716 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35647 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3464 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700499325 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701187862 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10708837 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708851 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708845 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708847 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708839 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708841 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708844 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708846 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708842 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708850 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708843 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708835 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708840 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708838 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708848 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10708834 | - | - |
Rat Genome 230 2.0 Array | 1389093_PM_at | - | - |
Rat Genome 230 2.0 Array | 1387377_a_at | - | - |
Rat Genome 230 2.0 Array | 1389093_at | - | - |
Rat Genome 230 2.0 Array | 1387377_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1387377_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1387377_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100016049.rn.1 | TC0100002088.rn.1 | RTA TC |
Rat Expression Set 230 | 1387377_PM_a_at | - | - |
Rat Expression Set 230 | 1389093_at | - | - |
Rat Expression Set 230 | 1387377_a_at | - | - |
Rat Expression Set 230 | 1389093_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10708834 | - | - |
Rat Genome U34 Set | U49953_s_at | - | - |
Rat Genome U34 Set | rc_AI172085_at | - | - |
Rat Genome U34 Set | rc_AI175989_at | - | - |
Rat Genome U34 Set | AA851136_g_at | - | - |
Rat Genome U34 Set | U49953_s_at | - | - |
Rat Genome U34 Set | AA851136_g_at | - | - |
Rat Genome U34 Set | U49953_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20915766 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362113 | - | - |
RatRef-12 Expression BeadChip | ILMN_69048 | - | - |
