Assay Details
| Gene Symbol: Fxyd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022388.1 | NP_071783.1 | 3 - 4 | 993 | 93 | |
| XM_006237206.2 | XP_006237268.1 | 4 - 5 | 332 | 93 | ||
| XM_008763260.1 | XP_008761482.1 | 4 - 5 | 662 | 93 | ||
| XM_008763261.1 | XP_008761483.1 | 4 - 5 | 620 | 93 | ||
| GenBank mRNA | L41254.1 | - | 3 - 4 | 993 | 93 | |
More Information
Set Membership: |
|

Fxyd4
Genomic Map