Assay Details
Gene Symbol: Pip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022708.2 | NP_073199.1 | 1 - 2 | 100 | 102 | |
GenBank mRNA | AF054270.1 | - | 1 - 2 | 100 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
