Assay Details
Gene Symbol: Nolc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022869.1 | NP_074060.1 | 7 - 8 | 940 | 80 | |
XM_006231560.2 | XP_006231622.1 | 7 - 8 | 988 | 80 | ||
GenBank mRNA | BC126088.1 | - | 7 - 8 | 905 | 80 | |
M94287.1 | - | 7 - 8 | 940 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
